Learn More
Merck Accessories for YY3014236 Filter Holder, 142 mm
Accessories for filter holders for sample preparation.
Brand: Merck YY3014234
700.85 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Filter diameter: 142mm
Specifications
Not Autoclavable | |
142 mm filter holders | |
Non-sterile |
Support Screen | |
PTFE Coated 316 Stainless Steel | |
Laboratory Filter Holders or Cyclones |