Learn More
Merck Stainless Steel Plug, 1/4 in. NPT
Shop All Merck Products
Click to view available options
:
Plug, 1/4 in. NPT, stainless steel
Description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel. Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Dispensing Pressure Vessel |
| Material | Stainless Steel |
| Sterility | Non-sterile |
| Type | Plug |
| Diameter (Metric) Inside | 2.5 mm |