Learn More
Merck MultiScreen™ Column Loaders
Shop All Merck Products
Click to view available options
Volume (Metric) Well:
25 μL
45 μL
100 μL
Unit Size:
Each
Description
- Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
Specifications
Specifications
| Color | Black |
| Product Type | MultiScreen Column Loader |
| Volume (Metric) Well | 100 μL |
| Sterility | Non-sterile |
| For Use With (Application) | Load 100μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography |
| Quantity | 1/Pk. |