Learn More
Merck MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates
Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model
Brand: Merck MANMN2010
466.41 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.
- Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
- Optically clear for use with an imaging microscope or other microscopic analysis
- Can be coated with a thin layer of agar if necessary
- Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
0.28 cm2 | |
10/Pk. |
Nylon Mesh | |
20 μm | |
Styrene | |
Non-sterile | |
Whole organism screening |