missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Petri Dishes and Petri-Pad™ Dishes

Available with or without absorbent pads

Brand:  Merck Millipore PD20047S0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11903876

  • € 138.00 / Pack of 150


Estimated Shipment: 23-07-2021
Add to basket

Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 045 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)

Description

Description

Available with or without absorbent pads

Petri Dishes

  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose

Specifications

Specifications

Clear
47 mm
Polystyrene
Disposable
150 Pack
Round
Sterile
Dish with Pad
Sleeve
Videos
SDS
Documents
Certificates