Learn More
Merck Replacement Parts for Stainless-Steel 13 and 25mm Filter Holders
Use to ultraclean or sterilize small volumes of liquids dispensed by syringe
Brand: Merck XX3001202
105.40 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Swinnex™ parts kit contains two PTFE gaskets, two O-rings, and two support screens.
Specifications
Stainless-Steel Filter Holder Replacement Parts | |
PTFE | |
Flat gasket |