missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emedastine Difumarate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Emedastine difumarate |
| CAS | 87233-62-3 |
| Analyte or Component Names | Emedastine difumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H26 N4 O . 2 C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Emedastine difumarate, 1-(2-Ethoxyethyl)-2-(4-methylhexahydro-1H-1,4-diazepin-1-yl)-1H-benzimidazole bis[hydrogen (2E)-butenedioate], 1H-Benzimidazole, 1-(2-ethoxyethyl)-2-(hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-, (2E)-2-butenedioate (1:2), 1H-Benzimidazole, 1-(2-ethoxyethyl)-2-(hexahydro-4-methyl-1H-1,4-diazepin-1-yl)-, (E)-2-butenedioate (1:2), 1H-1,4-Diazepine, 1H-benzimidazole deriv., AL 3432A, Emadine, KB 2413, KG 2413, LY 188695, Rapimine |
| InChI Formula | InChI=1S/C17H26N4O.2C4H4O4/c1-3-22-14-13-21-16-8-5-4-7-15(16)18-17(21)20-10-6-9-19(2)11-12-20;2*5-3(6)1-2-4(7)8/h4-5,7-8H,3,6,9-14H2,1-2H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1+ |
| Solution Type | Neat |
| SMILES | CCOCCn1c(nc2ccccc12)N3CCCN(C)CC3.OC(=O)\C=C\C(=O)O.OC(=O)\C=C\C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?