missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMG1 Antibody (OTI1B8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71601B
This item is not returnable.
View return policy
Description
EMG1 Monoclonal specifically detects EMG1 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| EMG1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| EMG1 | |
| Full length recombinant protein of human EMG1 (NP_006322) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1B8 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1, BWCNS, C2F18S rRNA Psi1248 methyltransferase, EMG1 nucleolar protein homolog (S. cerevisiae), essential for mitotic growth 1, FLJ60792, Grcc2f, NEP1EC 2.1.1.-, Nucleolar protein EMG1 homolog, probable ribosome biogenesis protein NEP1, Protein C2f, Ribosome biogenesis protein NEP1 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 10436.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction