missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11965-1-AP-20UL
This item is not returnable.
View return policy
Description
EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.Specifications
| EMG1 | |
| Polyclonal | |
| Unconjugated | |
| EMG1 | |
| C2F, EMG1, Grcc2f, NEP1, Nucleolar protein EMG1 homolog, Protein C2f | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10436, 14791, 312706 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35130, Q92979 | |
| EMG1 | |
| EMG1 Fusion Protein Ag2575 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction