missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMI1 Antibody (EMI1/1176), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47786F
This item is not returnable.
View return policy
Description
EMI1 Monoclonal antibody specifically detects EMI1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| EMI1 | |
| Monoclonal | |
| FITC | |
| EMI1Early mitotic inhibitor 1, F-box only protein 5, F-box protein 5, FBX5F-box protein Fbx5, Fbxo31 | |
| Recombinant fragment (203 amino acid residues between aa 1-250) of human EMI1 protein (Uniprot: Q9UKT4) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| EMI1/1176 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 26271 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction