missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMI1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77748
This item is not returnable.
View return policy
Description
EMI1 Polyclonal specifically detects EMI1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EMI1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EMI1Early mitotic inhibitor 1, F-box only protein 5, F-box protein 5, FBX5F-box protein Fbx5, Fbxo31 | |
| Rabbit | |
| 50.15 kDa | |
| 0.1 mg | |
| Primary | |
| This antibody is directed against human Fbp5A protein. A BLAST analysis was used to suggest cross reactivity with Fbp5A protein. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.3 mg/ml | |
| Western Blot 1:500-1:3000, ELISA 1:5000-1:20000, Immunohistochemistry 1:500-1:3000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9UK58 | |
| FBXO5 | |
| This affinity purified EMI1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the amino terminal end of human EMI1 protein. (Uniprot: Q9UKT4) | |
| Affinity Purified | |
| RUO | |
| 26271 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction