missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EML4 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05974B
This item is not returnable.
View return policy
Description
EML4 Polyclonal antibody specifically detects EML4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EML4 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C2orf2, echinoderm microtubule associated protein like 4, echinoderm microtubule-associated protein-like 4, ELP120, EMAP-4, EMAPL4, FLJ10942, FLJ32318, Restrictedly overexpressed proliferation-associated protein, Ropp 120, ROPP120DKFZp686P18118 | |
| The immunogen recognized by this antibody maps to a region between residue 931 and 981of human echinoderm microtubule associated protein like 4 using the numbering given in entry NP_061936.2 (GeneID 27436). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 27436 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction