missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EML4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15635105
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15635105 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15635105 Supplier Invitrogen™ Supplier No. PA551181

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total EML4 protein.

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT, interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene EML4
Gene Accession No. Q9HC35
Gene Alias 4930443C24Rik; AI644019; C2orf2; echinoderm microtubule associated protein like 4; echinoderm microtubule-associated protein-like 4; ELP120; EMAP-4; EMAPL4; EML4; OTTHUMP00000201447; OTTHUMP00000201455; Restrictedly overexpressed proliferation-associated protein; ropp 120; ROPP120
Gene Symbols EML4
Host Species Rabbit
Immunogen Full length fusion protein of human EML4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27436
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.