missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMMPRIN/CD147 Antibody (1159A), PE/Cy7, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB7721PECY7
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Flow Cytometry
Specifications
| EMMPRIN/CD147 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1159A | |
| Flow Cytometry | |
| 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| NS0 mouse myeloma cell line transfected with mouse EMMPRIN/CD147, Ala22-Pro209, Accession # P18572 | |
| 0.1 mL | |
| Cancer, Signal Transduction, Vision | |
| 682 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction