Learn More
EMMPRIN/CD147 Antibody (125), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89320V
Product Code. 18269488
  • ONLINE EXCLUSIVE PRICE
    € 649.00
    € 613.20 / 0.10mL
    Save € 35.80
    5% Off
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EMMPRIN/CD147
Monoclonal
DyLight 405
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
This antibody was obtained from a rabbit immunized with purified, recombinant Human EMMPRIN/CD147 (NP_940991.1; Met 1-His 205).
0.1 mL
Cancer, Signal Transduction, Vision
682
Store at 4°C in the dark.
IgG
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
125
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human, Monkey