missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06088MFV450
This item is not returnable.
View return policy
Description
EMR1 Polyclonal antibody specifically detects EMR1 in Human samples. It is validated for Western Blot
Specifications
| EMR1 | |
| Polyclonal | |
| Western Blot | |
| ADGRE1, egf-like module containing, mucin-like, hormone receptor-like 1, egf-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EMR1 hormone receptor, TM7LN3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human EMR1. | |
| 0.1 mL | |
| Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
| 2015 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction