missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR2 Antibody (494025), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4894MFV610
This item is not returnable.
View return policy
Description
EMR2 Monoclonal antibody specifically detects EMR2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| EMR2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 30817 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 494025 | |
| Flow Cytometry, CyTOF-ready | |
| CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2 | |
| NS0 mouse myeloma cell line transfected with human EMR2, Gln24-Asn823, Accession # AAI27006 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction