missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMR2 Polyclonal Antibody

EMR2 Polyclonal Antibody
Brand: Invitrogen PA5102055
This item is not returnable.
View return policy
Description
EMR2 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| EMR2 | |
| Polyclonal | |
| Unconjugated | |
| ADGRE2 | |
| ADGRE2; Adhesion G protein-coupled receptor E2; CD312; CD312 antigen; egf-like module containing, mucin-like, hormone receptor-like 2; EGF-like module receptor 2; EGF-like module-containing mucin-like hormone receptor-like 2; EMR2; VBU | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 30817 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UHX3 | |
| ADGRE2 | |
| A synthesized peptide derived from human ADGRE2(Accession Q9UHX3), corresponding to amino acid residues R128-T178. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction