missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMSY Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-545AF594
This item is not returnable.
View return policy
Description
EMSY Polyclonal antibody specifically detects EMSY in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EMSY | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| chromosome 11 open reading frame 30, EMSYFLJ90741, protein EMSY | |
| The immunogen recognized by this antibody maps to a region between residue 1250 and the C-terminus (residue 1322) of human EMSY chromosome 11 open reading frame 30 using the numbering given in TrEMBL entry Q8TE50 (GeneID 56946). | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| 56946 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction