missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine, MedChemExpress

Product Code. 16417306
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16417306

Brand: Medchem Express HY17427

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Emtricitabine is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell. It is an antiviral drug for the treatment of HIV infection.

Emtricitabine is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell. It is an antiviral drug for the treatment of HIV infection.

Specifications

Chemical Name or Material Emtricitabine
Physical Form Solid
CAS 143491-57-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H10FN3O3S
For Use With (Application) COVID-19-anti-virus
Synonym BW1592
Solubility Information DMSO : ≥ 100 mg/mL (404.45 mM)
SMILES O=C1N=C(N)C(F)=CN1[C@@H]2CS[C@H](CO)O2
Molecular Weight (g/mol) 247.25
Formula Weight 247.25
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.