missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EMX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552294
This item is not returnable.
View return policy
Description
Immunogen sequence: RSLYGGPELV FPEAMNHPAL TVHPAHQLGA SPLQPPHSFF GAQHRDPLHF YPWVLRNRFF GHRFQASDVP QDGLLLHGPF ARKPKRI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.
EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.
Specifications
| EMX1 | |
| Polyclonal | |
| Unconjugated | |
| Emx1 | |
| empty spiracles homeobox 1; empty spiracles homolog 1; Empty spiracles-like protein 1; EMX1; Emx-1; Homeobox protein EMX1; RGD1564002 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13796, 2016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q04741, Q04742 | |
| Emx1 | |
| Recombinant protein fragment of Human EMX1 (Product #RP-89351) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction