missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EN 12918 Organophosphorus Mixture 439 10 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Azinphos-ethyl∣Azinphos-methyl∣Bromophos-methyl∣Chlorfenvinphos∣Chlorpyrifos∣Chlorpyrifos-methyl∣Diazinon∣Dichlorvos∣Dimethoate∣Fenitrothion∣Fenthion∣Malathion∣Mevinphos∣Parathion-ethyl∣Parathion-methyl∣Phosalone∣Propetamphos∣Triadimefon∣Triazophos |
| CAS | 2642-71-9, 86-50-0, 2104-96-3, 470-90-6, 2921-88-2, 5598-13-0 |
| Analyte or Component Names | Azinphos-ethyl∣Azinphos-methyl∣Bromophos-methyl∣Chlorfenvinphos∣Chlorpyrifos∣Chlorpyrifos-methyl∣Diazinon∣Dichlorvos∣Dimethoate∣Fenitrothion∣Fenthion∣Malathion∣Mevinphos∣Parathion-ethyl∣Parathion-methyl∣Phosalone∣Propetamphos∣Triadimefon∣Triazophos |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 10 mL |
| Concentration or Composition (by Analyte or Components) | Azinphos-ethyl 10 μg/mL∣Azinphos-methyl 10 μg/mL∣Bromophos-methyl 10 μg/mL∣Chlorfenvinphos 10 μg/mL∣Chlorpyrifos 10 μg/mL∣Chlorpyrifos-methyl 10 μg/mL∣Diazinon 10 μg/mL∣Dichlorvos 10 μg/mL∣Dimethoate 10 μg/mL∣Fenitrothion 10 μg/mL∣Fenthion 10 μg/mL∣Malathion 10 μg/mL∣Mevinphos 10 μg/mL∣Parathion-ethyl 10 μg/mL∣Parathion-methyl 10 μg/mL∣Phosalone 10 μg/mL∣Propetamphos 10 μg/mL∣Triadimefon 10 μg/mL∣Triazophos 10 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?