Learn More
Justrite EN Cabinet Base Cover for 22607
Protect workers, reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets. Designed to meet OSHA and NFPA standards.
Brand: Justrite JCB22637YL
Additional Details : Weight : 1.80000kg
Description
- Protect workers, reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets
- Designed to meet OSHA and NFPA standards, Sure-Grip™ EX cabinets are constructed of sturdy 18 gauge (1 mm) thick double-wall, welded steel with 1 1/2 in. (38 mm) of insulating air space for fire resistance
- High-performance, self-latching doors close easily and securely for maximum protection under fire conditions
- Fail-safe closing mechanism ensures three-point stainless steel bullet latching system works every time
Specifications
Yellow | |
Base Cover | |
1.5 cm | |
111.8 cm |