missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EN-RAGE/S100A12 Antibody (001), DyLight 680 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89815FR
This item is not returnable.
View return policy
Description
EN-RAGE/S100A12 Monoclonal antibody specifically detects EN-RAGE/S100A12 in Human samples. It is validated for ELISA
Specifications
| EN-RAGE/S100A12 | |
| Monoclonal | |
| DyLight 680 | |
| CAAF1Neutrophil S100 protein, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EN-RAGE/S100A12 (NP_005612.1; Met1-Glu92). | |
| 0.1 mL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 6283 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 001 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction