missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EN-RAGE/S100A12 Antibody (OTI1D1), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71276C
This item is not returnable.
View return policy
Description
EN-RAGE/S100A12 Monoclonal specifically detects EN-RAGE/S100A12 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EN-RAGE/S100A12 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| S100A12 | |
| Full length recombinant protein of human EN-RAGE/S100A12 (NP_005612) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1D1 | |
| Western Blot, Immunohistochemistry | |
| CAAF1Neutrophil S100 protein, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C) | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 6283.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction