missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EN1/Engrailed 1 Antibody (SR1987), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | EN1/Engrailed 1 |
|---|---|
| Clone | SR1987 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682444
|
Novus Biologicals
NBP3-22202-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672275
|
Novus Biologicals
NBP3-22202-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EN1/Engrailed 1 Monoclonal antibody specifically detects EN1/Engrailed 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
| EN1/Engrailed 1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| engrailed homeobox 1, engrailed homolog 1, Homeobox protein en-1, homeobox protein engrailed-1, hu-En-1 | |
| A synthesized peptide derived from human EN1/Engrailed 1 (Uniprot #: Q05925) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1987 | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience, Stem Cells | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 2019 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title