missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enalapril Ethyl Ester (N-[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-L-proline Ethyl Ester), Mikromol™
Description
Enalapril Ethyl Ester (N-[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-L-proline Ethyl Ester)
Specifications
Specifications
| Chemical Name or Material | Enalapril Ethyl Ester |
| CAS | 87420-38-0 |
| Analyte or Component Names | Enalapril Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H32 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-L-proline ethyl ester (9CI) |
| InChI Formula | InChI=1S/C22H32N2O5/c1-4-28-21(26)18(14-13-17-10-7-6-8-11-17)23-16(3)20(25)24-15-9-12-19(24)22(27)29-5-2/h6-8,10-11,16,18-19,23H,4-5,9,12-15H2,1-3H3/t16-,18-,19-/m0/s1 |
| SMILES | CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)OCC |
| IUPAC Name | ethyl (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?