Learn More
Enalaprilat-d5 sodium, MedChemExpress
Product Code. 16402109
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402109 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16402109 Supplier Medchem Express Supplier No. HYB0231BS
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
MedChemExpress Enalaprilat (MK-422) D5 Sodium Salt is the deuterium labeled Enalaprilat(MK-422), which is an angiotensin-converting enzyme (ACE) inhibitor.
Enalaprilat (MK-422) D5 Sodium Salt is the deuterium labeled Enalaprilat(MK-422), which is an angiotensin-converting enzyme (ACE) inhibitor.
Specifications
Chemical Name or Material Enalaprilat-d5 sodium
CAS 1356922-29-6
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C18H17D5N2Na2O5
Color White
Synonym MK-422-d5 sodium
Solubility Information H2O : ≥ 100 mg/mL (251.64 mM)
SMILES C[C@@H](C(N1CCC[C@H]1C(O[Na])=O)=O)N[C@H](C(O[Na])=O)CCC2=C([2H])C([2H])=C([2H])C([2H])=C2[2H]
Industry Type COVID-19-anti-virus
Molecular Weight (g/mol) 397.39
Quantity 1 mg
Formula Weight 397.39
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only