Learn More
Enantiomer of Sofosbuvir, MedChemExpress
Product Code. 16442469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442469 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16442469 Supplier Medchem Express Supplier No. HYI0726
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Enantiomer of Sofosbuvir is an enantiomer of Sofosbuvir, a prescription medicine for the treatment of patients with chronic hepatitis C. There is no biological activity report on enantiomer of Sofosbuvir until now.
Enantiomer of Sofosbuvir is an enantiomer of Sofosbuvir, a prescription medicine for the treatment of patients with chronic hepatitis C. There is no biological activity report on enantiomer of Sofosbuvir until now.
Specifications
Chemical Name or Material Enantiomer of Sofosbuvir
Color White
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 1 mg
SMILES O=[P@](N[C@H](C)C(OC(C)C)=O)(OC[C@H]1[C@H](O)[C@@](F)(C)[C@@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only