missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endo G Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 359.00 - € 515.00
Specifications
| Antigen | Endo G |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Endo G Polyclonal antibody specifically detects Endo G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Endo G | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Hypoxia | |
| PBS (pH 7.2) and 40% Glycerol | |
| 2021 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.30.-, Endo G, endonuclease G, endonuclease G, mitochondrial, FLJ27463, mitochondrial endonuclease G | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QVIGKNHVAVPTHFFKVLILEAAGGQIELRTYVMPNAPVDEAIPLERFLVPIESIERASGLLFVPNILARAGSLKAITAGSK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title