missing translation for 'onlineSavingsMsg'
Learn More

Endo G Antibody (OTI5F1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-70645AF594

Product Code. 18451364

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Endo G Monoclonal antibody specifically detects Endo G in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endo G
Monoclonal
Alexa Fluor 594
EC 3.1.30.-, Endo G, endonuclease G, endonuclease G, mitochondrial, FLJ27463, mitochondrial endonuclease G
Full length recombinant protein of human Endo G (NP_004426) produced in HEK293T cells.
0.1 mL
Apoptosis, Cancer, Hypoxia
2021
Store at 4°C in the dark.
IgG1
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
OTI5F1
50mM Sodium Borate
Mouse
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.