missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endocan/ESM-1 Antibody (RM0079-5J44), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21710C
This item is not returnable.
View return policy
Description
Endocan/ESM-1 Monoclonal specifically detects Endocan/ESM-1 in Mouse samples. It is validated for Western Blot.
Specifications
| Endocan/ESM-1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ESM1 | |
| Mouse recombinant Endocan | |
| 0.1 mL | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Endocan. | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0079-5J44 | |
| Western Blot | |
| endocan, endothelial cell-specific molecule 1, ESM-1 | |
| Rat | |
| Protein A or G purified | |
| Angiogenesis, Cancer | |
| 11082 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction