missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Antibody (MEM-226), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-452AF647
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Flow (Cell Surface).
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-226 reacts with CD105 (Endoglin), a 90 kDa type I homodimerizing membrane glycoprotein expressed on vascular endothelial cells (small and large vessels), activated monocytes and tissue macrophages, stromal cells of certain tissues including bone marrow, pre-B lymphocytes in fetal marrow and erythroid precursors in fetal and adult bone marrow; it is also present on syncytiotrophoblast on placenta throughout pregnancy. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry | |
| MEM-226 | |
| Western Blot, Flow Cytometry, Flow (Cell Surface) | |
| ENG | |
| Recombinant full length protein (Human). Expressed in vaccinia virus containing CD105 cDNA. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2022 | |
| Human, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction