missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Endoglin/CD105 Antibody Pair [HRP]

Product Code. 18612570 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18612570

Brand: Novus Biologicals™ NBP2794525Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Endoglin/CD105.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 15.63-1000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Endoglin/CD105.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Gene ID (Entrez) 2022
Gene Symbol ENG
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 120 ng of recombinant Endoglin/CD105.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Endoglin/CD105 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL mouse anti-Endoglin/CD105 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Endoglin/CD105
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.