missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglycan/PODXL2 Antibody (MM0258- 7R2), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12273AF594
This item is not returnable.
View return policy
Description
Endoglycan/PODXL2 Monoclonal specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot.
Specifications
| Endoglycan/PODXL2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PODXL2 | |
| Human recombinant Endoglycan/PODXL2 (extracellular domain) | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot | |
| MM0258- 7R2 | |
| Western Blot | |
| endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2 | |
| Mouse | |
| 0.1 mL | |
| 50512 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction