Learn More
Invitrogen™ Endomucin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597084
Description
Immunogen sequence: NSTGVLEAAN NSLVVTTTKP SITTPNTESL QKNVVTPTTG TTPKGTITNE LLKMSLMSTA TFLTSKDEGL KATTTDVRKN DSIISNVTVT SVTLPNAVST LQSSKPKTET QSSIKTT; Positive Samples: MCF-7, NIH/3T3; Cellular Location: Cell membrane, Membrane, Single-pass type I membrane protein, Single-pass type I membrane protein.
Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.
Specifications
| Endomucin | |
| Polyclonal | |
| Unconjugated | |
| Emcn | |
| 0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51705, 59308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9R0H2, Q9ULC0 | |
| Emcn | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-135 of human Endomucin (NP_0573262). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.