missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Endomucin Recombinant Rabbit Monoclonal Antibody (4O5S7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558144
This item is not returnable.
View return policy
Description
Immunogen sequence: LYRMCWKADP GTPENGNDQP QSDKESVKLL TVKTISHESG EHSAQGKTKN.
Endomucin (EMCN) also known as endothelial sialomucin or mucin-like sialoglycoprotein interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.
Specifications
| Endomucin | |
| Recombinant Monoclonal | |
| 1.85 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9R0H2 | |
| Emcn | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 212-261 of human Endomucin (NP_057326.2). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 4O5S7 | |
| Unconjugated | |
| Emcn | |
| 0610012K22Rik; AI315669; Emcn; EMCN2; Endomucin; endomucin-1; endomucin-1/2; Endomucin-2; gastric cancer antigen Ga34; Muc14; MUC-14; mucin 14, endothelial; mucin-14; sialomucin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 59308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction