missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Antibody (SPM255), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-11461DL594
This item is not returnable.
View return policy
Description
Endorepellin/Perlecan/Heparan Sulfate Proteoglycan Monoclonal antibody specifically detects Endorepellin/Perlecan/Heparan Sulfate Proteoglycan in Human, Mouse, Porcine, Bovine, Fish, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Endorepellin/Perlecan/Heparan Sulfate Proteoglycan | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bovine, Fish, Monkey | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM255 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 | |
| Murine EHS laminin preparation | |
| 0.1 mL | |
| Cell Biology | |
| 3339 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction