missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Endosialin/CD248/TEM1 Antibody (ontuxizumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28072-50ug
This item is not returnable.
View return policy
Description
Endosialin/CD248/TEM1 Monoclonal antibody specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Endosialin/CD248/TEM1 | |
| Monoclonal | |
| Unconjugated | |
| Q9HCU0 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ontuxizumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1 | |
| TEM1/ Endosialin/ CD248 | |
| 50 μg | |
| Cancer | |
| 57124 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction