missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endostatin Antibody (RM0060-3H11), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12274G
This item is not returnable.
View return policy
Description
Endostatin Monoclonal specifically detects Endostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Endostatin | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| COL18A1 | |
| Mouse recombinant Endostatin | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0060-3H11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP | |
| Rat | |
| Protein A or G purified | |
| Angiogenesis, Biologically Active Proteins, Cancer, Extracellular Matrix, Tumor Suppressors | |
| 80781 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction