missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelial Lipase Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-118AF700
This item is not returnable.
View return policy
Description
Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Endothelial Lipase | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Q9Y5X9 | |
| LIPG | |
| An N-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9] | |
| 0.1 mL | |
| Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Unfolded Protein Response | |
| 9388 | |
| Human, Mouse (Negative) | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Does not appear to recognize mouse. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction