missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelial Lipase Antibody (OTI1D9), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71309MFV500
This item is not returnable.
View return policy
Description
Endothelial Lipase Monoclonal antibody specifically detects Endothelial Lipase in Human samples. It is validated for Flow Cytometry
Specifications
| Endothelial Lipase | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| OTI1D9 | |
| Flow Cytometry | |
| EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719 | |
| Full length human recombinant protein of human LIPG (NP_006024) produced in HEK293T cell. | |
| 0.1 mL | |
| Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Microautophagy, Unfolded Protein Response | |
| 9388 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction