missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33578
This item is not returnable.
View return policy
Description
Endothelin-3 Polyclonal specifically detects Endothelin-3 in Human samples. It is validated for Western Blot.
Specifications
| Endothelin-3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EDN3 | |
| Recombinant protein encompassing a sequence within the center region of human Endothelin 3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 1908 | |
| Human | |
| IgG |
| Western Blot | |
| 0.52mg/mL | |
| Western Blot 1:500-1:3000 | |
| endothelin 3, endothelin-3, ET3, ET-3, HSCR4, MGC15067, MGC61498, PPET3, preproendothelin-3, truncated endothelin 3, WS4B | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction