Learn More
Endothelin 3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27871-1-AP-20UL
Description
The Endothelin 3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Endothelin 3. This antibody recognizes human, mouse, rat antigen. The Endothelin 3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Endothelin 3 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| EDN3, endothelin 3, ET 3, ET3, PPET3, Preproendothelin 3 | |
| 20 μL | |
| 1908 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |