Learn More
Endothelin 3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27871-1-AP-150UL
 View more versions of this product
Product Code. 18098955
  • € 410.00 / 150µL
Description
The Endothelin 3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Endothelin 3. This antibody recognizes human, mouse, rat antigen. The Endothelin 3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Endothelin 3
550 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
EDN3, endothelin 3, ET 3, ET3, PPET3, Preproendothelin 3
150 μL
1908
Store at -20°C. Stable for one year after shipment.