missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelium Monoclonal Antibody (OX-43), Biotin
Endothelium Monoclonal Antibody (OX-43), Biotin
Brand: Invitrogen MA517515
This item is not returnable.
View return policy
Description
Specifications
| Endothelium | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| OX-43 | |
| Biotin | |
| Endothelial cell marker; Endothelial cells; Endothelium Cell Antigen-1; Pan endothelial cells | |
| Rat Peritoneal Macrophages. | |
| 500 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction