missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENG Monoclonal Antibody (OTI6G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506144
This item is not returnable.
View return policy
Description
CD105 (Endoglin is a Type I transmembrane protein) is highly expressed on human vascular endothelial cells and exists on an O- and N-glycosylated homodimer. Up regulation of CD105 expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 is highly expressed on endothelial cells and promotes angiogenesis during wound healing, infarcts and in a wide range of tumors and its gene expression is stimulated by hypoxia. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2. Mutations in the CD105 gene cause hereditary hemorrhagic telangiectasia, also known as Osler-Rendu-Weber syndrome 1, an autosomal domit multisystemic vascular dysplasia. Alternatively spliced transcript variants encoding different isoforms of CD105 have been found. Diseases associated with CD105 dysfunction include hereditary hemorrhagic telangiectasia.Specifications
| ENG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17813 | |
| ENG | |
| Full length human recombit protein of human ENg produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI6G8 | |
| Unconjugated | |
| ENG | |
| END, HHT1, ORW1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2022 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction