Learn More
Karl Hecht Narrow-necked bottleswithglass stoppers, amber glass 2000ml
Brand:  Karl Hecht 41303036
Narrow-necked bottleswithglass stoppers, amber glass 2000ml
Product Code. 10093741
  • ONLINE EXCLUSIVE PRICE
    € 23.10
    € 21.74 / Each
    Save € 1.36
    5% Off
Only null left
Estimated Shipment: 15-01-2026