Learn More
Invitrogen™ Enlimomab Recombinant Mouse Monoclonal Antibody
GREENER_CHOICE
Mouse Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA541744
Product Code. 17768363
  • € 500.00 / 100µg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Enlimomab is a biosimilar that targets ICAM1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Enlimomab
Recombinant Monoclonal
Unconjugated
BI-RR-1; BIRR-1
Human ICAM1.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
0.61 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein A/G
RUO
Human
Antibody
IgG2a