missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENO1, Mouse anti-Human, Clone: 1G1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00002023-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: MSILKIHAREIFDSRGNPTVEVDLFTSKGLFRAAVPSGASTGIYEALELRDNDKTRYMGKGVSKAVEHINKTIAPALVSKKLNVTEQEKIDKLMIEMDGTENKSKFGANAILGVSLAVCKAGAVEKGVPLYRHIADLAGNSEVILPVPAFNVINGGSHAGNKLAMQEFMILPVGAANFREAMRIGAEVYHNLKNVIKEKYGKDATNVGDEGGFAPNILENKEGLELLKTAIGKAGYTDKVVIGMDVAASEFFRSGKYDLDFKSPDDPSRYISPDQLADLYKSFIKDYPVVSIEDPFDQDDWGAWQKFTASAGIQVVGDDLTVTNPKRIAKAVNEKSCNCLLLKVNQIGSVTESLQACKLAQANGWGVMVSHRSGETEDTFIADLVVGLCTGQIKTGAPCRSERLAKYNQLLRIEEELGSKAKFAGRNFRNPLAKSpecifications
| ENO1 | |
| Monoclonal | |
| Unconjugated | |
| enolase 1, (alpha) | |
| ENO1L1/MBP-1/MPB1/NNE/PPH | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
| ELISA, Western Blot | |
| 1G1 | |
| In 1x PBS, pH 7.4 | |
| BC022545 | |
| ENO1 | |
| ENO1 (AAH22545, 1 a.a. ∼ 434 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 2023 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction